Assay Details
Target Gene Details
Entrez Gene ID: | 4521 |
Gene Name: | nudix hydrolase 1 |
Gene Aliases: |
MTH1 |
Location: |
Chr.7:2242222-2251145 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDT1 | NM_002452.3 | 4 | 470 | NP_002443.3 |
| NM_198948.1 | 5 | 521 | NP_945186.1 | |
| NM_198949.1 | 5 | 594 | NP_945187.1 | |
| NM_198950.1 | 5 | 518 | NP_945188.1 | |
| NM_198952.1 | 5 | 591 | NP_945190.1 | |
| NM_198953.1 | 4 | 506 | NP_945191.1 | |
| NM_198954.1 | 4 | 579 | NP_945192.1 | |
| AB025233.1 | 4 | 470 | BAA83791.1 | |
| AB025234.1 | 5 | 502 | BAA83792.1 | |
| AB025235.1 | 5 | 575 | BAA83793.1 | |
| AB025236.1 | 5 | 518 | BAA83794.1 | |
| AB025237.1 | 5 | 591 | BAA83795.1 | |
| AB025238.1 | 4 | 506 | BAA83796.1 | |
| AB025239.1 | 4 | 579 | BAA83797.1 | |
| AB025240.1 | 5 | 575 | BAA83798.1 | |
| AB025241.1 | 5 | 591 | BAA83799.1 | |
| AB025242.1 | 4 | 579 | BAA83800.1 | |
| AK026631.1 | 5 | 519 | ||
| BC014618.2 | 4 | 573 | AAH14618.1 | |
| BC040144.1 | 4 | 481 | ||
| BC051375.1 | 4 | 456 | ||
| BC065367.1 | 4 | 678 | AAH65367.1 | |
| BM927392.1 | 4 | 677 | ||
| CR407655.1 | 3 | 423 | CAG28583.1 | |
| D16581.1 | 3 | 449 | BAA04013.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1021310 | Chr.7:1936627 - 2280155 on Build GRCh38 | Loss |
|
| nsv1018563 | Chr.7:2233301 - 2325297 on Build GRCh38 | Gain |
|
| nsv1021173 | Chr.7:2206456 - 2334992 on Build GRCh38 | Loss |
|
| nsv830886 | Chr.7:2145358 - 2337903 on Build GRCh38 | Loss |
|
| esv2761136 | Chr.7:2230260 - 2325309 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map