Assay Details
Target Gene Details
Entrez Gene ID: | 23245 |
Gene Name: | astrotactin 2 |
Gene Aliases: |
bA67K19.1 |
Location: |
Chr.9:116425225-117415039 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASTN2 | NM_014010.4 | NP_054729.3 | ||
| AB014534.1 | ||||
| AF116574.1 | AAF14357.1 | |||
| AK127350.1 | ||||
| AK315025.1 | ||||
| BC018759.2 | AAH18759.2 | |||
| BC146756.1 | ||||
| BX649176.1 |
Target Gene Details
Entrez Gene ID: | 22954 |
Gene Name: | tripartite motif containing 32 |
Gene Aliases: |
BBS11, HT2A, LGMD2H, TATIP |
Location: |
Chr.9:116687302-116701300 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIM32 | NM_001099679.1 | 2 | 2651 | NP_001093149.1 |
| NM_012210.3 | 2 | 2654 | NP_036342.2 | |
| XM_005251813.3 | 2 | 2700 | XP_005251870.1 | |
| XM_011518398.2 | 2 | 2688 | XP_011516700.1 | |
| XM_017014486.1 | 3 | 2711 | XP_016869975.1 | |
| AX775779.1 | 2 | 2623 | CAE11589.1 | |
| BC003154.1 | 2 | 2623 | AAH03154.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047432 | Chr.9:116633553 - 116739347 on Build GRCh38 | Loss |
|
| dgv7708n100 | Chr.9:116663365 - 116741547 on Build GRCh38 | Loss |
|
| nsv615255 | Chr.9:116695698 - 116728293 on Build GRCh38 | Loss |
|
| nsv831702 | Chr.9:116638034 - 116816486 on Build GRCh38 | Gain |
|
| nsv1047284 | Chr.9:116639065 - 116863135 on Build GRCh38 | Loss |
|
| nsv1043149 | Chr.9:116677781 - 116825537 on Build GRCh38 | Loss |
|
| esv3573372 | Chr.9:116559083 - 116722034 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map