Assay Details
Target Gene Details
Entrez Gene ID: | 81618 |
Gene Name: | integral membrane protein 2C |
Gene Aliases: |
BRI3, BRICD2C, E25, E25C, ITM3 |
Location: |
Chr.2:230864598-230879254 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITM2C | NM_001012514.2 | 1 | 176 | NP_001012532.1 |
| NM_001012516.2 | 1 | 121 | NP_001012534.1 | |
| NM_001287240.1 | NP_001274169.1 | |||
| NM_001287241.1 | 2 | 308 | NP_001274170.1 | |
| NM_030926.5 | 1 | 176 | NP_112188.1 | |
| AB003629.1 | BAB46927.1 | |||
| AF271781.1 | 1 | 161 | AAG44792.1 | |
| AF272043.1 | 1 | 158 | AAF89492.1 | |
| AK056321.1 | 1 | 123 | ||
| AK075361.1 | 1 | 141 | BAC11570.1 | |
| AK090975.1 | BAC03562.1 | |||
| AK222697.1 | 1 | 123 | BAD96417.1 | |
| AK295503.1 | 1 | 89 | ||
| AK303199.1 | 1 | 141 | ||
| AK307702.1 | 1 | 116 | ||
| AL136603.1 | 1 | 117 | CAB66538.1 | |
| AL713651.1 | 1 | 100 | CAD28460.1 | |
| AY049777.1 | 1 | 62 | AAL15434.1 | |
| BC002424.1 | 1 | 103 | AAH02424.1 | |
| BC025742.1 | 1 | 91 | AAH25742.1 | |
| BC050668.1 | 1 | 111 | AAH50668.1 | |
| BC098563.1 | 1 | 89 | AAH98563.1 | |
| BM842614.1 | 1 | 165 | ||
| BX444331.2 | ||||
| CR457208.1 | CAG33489.1 | |||
| DA138409.1 | 2 | 145 | ||
| DA182471.1 | 1 | 176 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834565 | Chr.2:230697957 - 230920661 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map