Assay Details
Target Gene Details
Entrez Gene ID: | 8187 |
Gene Name: | zinc finger protein 239 |
Gene Aliases: |
HOK-2, MOK2 |
Location: |
Chr.10:43554516-43574619 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
More Information
Set Membership: |
|

Genomic Map