Assay Details
Target Gene Details
Entrez Gene ID: | 55802 |
Gene Name: | decapping mRNA 1A |
Gene Aliases: |
HSA275986, Nbla00360, SMAD4IP1, SMIF |
Location: |
Chr.3:53283423-53347627 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCP1A | NM_001290204.1 | 6 | 1164 | NP_001277133.1 |
| NM_001290205.1 | 8 | 1333 | NP_001277134.1 | |
| NM_001290206.1 | 6 | 1150 | NP_001277135.1 | |
| NM_001290207.1 | 5 | 1036 | NP_001277136.1 | |
| NM_018403.6 | 7 | 1278 | NP_060873.4 | |
| AB075507.1 | 7 | 1194 | BAE45754.1 | |
| AJ275986.1 | 7 | 1261 | CAB77023.1 | |
| AK001969.1 | 7 | 1194 | BAA92008.1 | |
| AK295205.1 | 6 | 1062 | ||
| AY146651.1 | 7 | 1194 | AAN62763.1 | |
| BC007439.2 | 7 | 1169 | AAH07439.1 | |
| KU178716.1 | 7 | 1168 | ||
| KU178717.1 | 6 | 1054 | ||
| KU178718.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3825 | Chr.3:53286923 - 53293922 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map