Assay Details
Target Gene Details
Entrez Gene ID: | 9819 |
Gene Name: | TSC22 domain family member 2 |
Gene Aliases: |
TILZ4a, TILZ4b, TILZ4c |
Location: |
Chr.3:150408268-150460119 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSC22D2 | NM_001303264.1 | 1 | 2570 | NP_001290193.1 |
| NM_014779.3 | 1 | 2570 | NP_055594.1 | |
| NR_130136.1 | 1 | 2570 | ||
| XM_011513337.2 | 1 | 2637 | XP_011511639.1 | |
| AB014569.1 | 1 | 2570 | ||
| AK297383.1 | 2 | 464 | ||
| BC044643.1 | 1 | 1904 | AAH44643.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829755 | Chr.3:150274236 - 150438214 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map