Assay Details
Target Gene Details
Entrez Gene ID: | 83546 |
Gene Name: | retbindin |
Gene Aliases: |
- |
Location: |
Chr.19:12825477-12835428 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953977 | Chr.19:12666887 - 13095486 on Build GRCh38 | Deletion |
|

Genomic Map