Assay Details
Target Gene Details
Entrez Gene ID: | 10171 |
Gene Name: | RNA terminal phosphate cyclase like 1 |
Gene Aliases: |
RNAC, RPCL1 |
Location: |
Chr.9:4792834-4861077 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RCL1 | NM_001286699.1 | NP_001273628.1 | ||
| NM_001286700.1 | NP_001273629.1 | |||
| NM_001286701.1 | NP_001273630.1 | |||
| NM_005772.4 | NP_005763.3 | |||
| XM_006716715.3 | XP_006716778.1 | |||
| XM_011517673.2 | XP_011515975.1 | |||
| XM_017014176.1 | XP_016869665.1 | |||
| AF067172.1 | AAD32456.1 | |||
| AF161456.1 | AAF29016.1 | |||
| AJ276894.1 | CAB89811.1 | |||
| AK022904.1 | BAB14300.1 | |||
| AK225872.1 | ||||
| AL518076.3 | ||||
| AL560629.3 | ||||
| BC001025.2 | AAH01025.1 | |||
| EF553527.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6459 | Chr.9:4829645 - 4849963 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map