Assay Details
Target Gene Details
Entrez Gene ID: | 868 |
Gene Name: | Cbl proto-oncogene B |
Gene Aliases: |
Cbl-b, Nbla00127, RNF56 |
Location: |
Chr.3:105655461-105869552 on Build GRCh38 |
Assay Gene Location: | Within Exon 31 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829651 | Chr.3:105484580 - 105683292 on Build GRCh38 | Loss |
|
| nsv591242 | Chr.3:105654992 - 105751290 on Build GRCh38 | Loss |
|
| nsv525071 | Chr.3:105286009 - 105690477 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map