Assay Details
Target Gene Details
Entrez Gene ID: | 91368 |
Gene Name: | CDKN2A interacting protein N-terminal like |
Gene Aliases: |
- |
Location: |
Chr.5:134402065-134414625 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDKN2AIPNL | NM_080656.2 | NP_542387.1 | ||
| XM_011543709.2 | 2 | 416 | XP_011542011.1 | |
| BC008293.1 | AAH08293.1 | |||
| BC018086.1 | AAH18086.1 | |||
| DB480185.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs141449831] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map