Assay Details
Target Gene Details
Entrez Gene ID: | 50640 |
Gene Name: | patatin like phospholipase domain containing 8 |
Gene Aliases: |
IPLA2-2, IPLA2G, MMLA, PNPLA-gamma, iPLA2gamma |
Location: |
Chr.7:108470417-108528161 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PNPLA8 | NM_001256007.2 | 11 | 2841 | NP_001242936.1 |
| NM_001256008.2 | 10 | 2795 | NP_001242937.1 | |
| NM_001256009.2 | 10 | 2609 | NP_001242938.1 | |
| NM_001256010.2 | 10 | 2668 | NP_001242939.1 | |
| NM_001256011.2 | 11 | 2529 | NP_001242940.1 | |
| NM_015723.4 | 12 | 2953 | NP_056538.1 | |
| XM_005250396.4 | 11 | 3611 | XP_005250453.1 | |
| XM_011516274.2 | 11 | 3670 | XP_011514576.1 | |
| XM_011516275.2 | 11 | 3484 | XP_011514577.1 | |
| XM_017012261.1 | 10 | 3438 | XP_016867750.1 | |
| AB041261.1 | 10 | 2621 | BAA94997.1 | |
| AF217519.1 | 3 | 734 | AAF67630.1 | |
| AF263613.1 | 11 | 2647 | AAF75269.1 | |
| AK002115.1 | 7 | 1141 | BAA92090.1 | |
| AK291755.1 | 11 | 2658 | ||
| AL834147.1 | 10 | 2362 | CAD38859.1 | |
| BC032999.2 | 12 | 2829 | AAH32999.1 | |
| BX647865.1 | 10 | 2514 |
More Information
Set Membership: |
|

Genomic Map