Assay Details
Target Gene Details
Entrez Gene ID: | 27000 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C2 |
Gene Aliases: |
MPHOSPH11, MPP11, ZRF1, ZUO1 |
Location: |
Chr.7:103312474-103345362 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC2 | NM_001129887.1 | NP_001123359.1 | ||
| NM_014377.1 | NP_055192.1 | |||
| XM_005250269.2 | XP_005250326.1 | |||
| XM_011516028.2 | XP_011514330.1 | |||
| XM_011516029.2 | XP_011514331.1 | |||
| XM_011516030.2 | XP_011514332.1 | |||
| XM_017011968.1 | XP_016867457.1 | |||
| AK126382.1 | ||||
| AL832847.1 | ||||
| BC000859.1 | AAH00859.1 | |||
| BC125057.1 | ||||
| BC139751.1 | ||||
| X98260.1 | CAA66913.1 |
Target Gene Details
Entrez Gene ID: | 9512 |
Gene Name: | peptidase, mitochondrial processing beta subunit |
Gene Aliases: |
Beta-MPP, MPP11, MPPB, MPPP52, P-52 |
Location: |
Chr.7:103297412-103329902 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PMPCB | NM_004279.2 | 13 | 3806 | NP_004270.2 |
| XM_005250717.3 | XP_005250774.1 | |||
| XM_006716181.3 | XP_006716244.1 | |||
| AI261792.1 | 1 | 134 | ||
| AK297354.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831084 | Chr.7:103138461 - 103324523 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map