Assay Details
Target Gene Details
Entrez Gene ID: | 55809 |
Gene Name: | transcriptional regulating factor 1 |
Gene Aliases: |
BCAR2, HSA277276, RAPA, TREP132, TReP-132, dJ139D8.5 |
Location: |
Chr.6:42224933-42452216 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
More Information
Set Membership: |
|

Genomic Map