Assay Details
Target Gene Details
Entrez Gene ID: | 100506472 |
Gene Name: | uncharacterized LOC100506472 |
Gene Aliases: |
- |
Location: |
Chr.22:39432160-39437128 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100506472 | NR_040535.1 | 4 | 2976 |
Target Gene Details
Entrez Gene ID: | 10454 |
Gene Name: | TGF-beta activated kinase 1/MAP3K7 binding protein 1 |
Gene Aliases: |
3'-Tab1, MAP3K7IP1 |
Location: |
Chr.22:39399754-39437127 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAB1 | NM_153497.2 | 11 | 1405 | NP_705717.1 |
| AF425640.1 | 11 | 1374 | AAN32760.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471200 | Chr.22:39416404 - 39478309 on Build GRCh38 | Loss |
|
| nsv834199 | Chr.22:39324389 - 39531343 on Build GRCh38 | Loss |
|
| nsv3628 | Chr.22:39435098 - 39468570 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map