Assay Details
Target Gene Details
Entrez Gene ID: | 640 |
Gene Name: | BLK proto-oncogene, Src family tyrosine kinase |
Gene Aliases: |
MODY11 |
Location: |
Chr.8:11493991-11564599 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BLK | NM_001715.2 | 1 | 286 | NP_001706.2 |
| XM_011543824.1 | 1 | 307 | XP_011542126.1 | |
| XM_011543827.1 | 1 | 307 | XP_011542129.1 | |
| XM_011543828.2 | 1 | 307 | XP_011542130.1 | |
| XM_011543829.2 | 1 | 307 | XP_011542131.1 | |
| XM_017013730.1 | 1 | 307 | XP_016869219.1 | |
| BC018703.1 | 1 | 150 | ||
| BQ057058.1 | 1 | 286 | ||
| S76617.1 | 1 | 297 | AAB33265.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34097 | Chr.8:11319863 - 11632385 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map