Assay Details
Target Gene Details
Entrez Gene ID: | 4081 |
Gene Name: | mab-21 like 1 |
Gene Aliases: |
CAGR1, Nbla00126 |
Location: |
Chr.13:35473789-35476695 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAB21L1 | NM_005584.4 | 1 | 231 | NP_005575.1 |
| AB073388.1 | 1 | 227 | BAE45718.1 | |
| AB593021.1 | 1 | 170 | ||
| AB593022.1 | 1 | 225 | ||
| AK313185.1 | 1 | 225 | ||
| BC028170.1 | 1 | 158 | AAH28170.1 | |
| DA611205.1 | 1 | 231 | ||
| DA909813.1 | 1 | 229 |
Target Gene Details
Entrez Gene ID: | 26960 |
Gene Name: | neurobeachin |
Gene Aliases: |
BCL8B, LYST2 |
Location: |
Chr.13:34942287-35672737 on Build GRCh38 |
Assay Gene Location: | Within Intron 43 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NBEA | NM_015678.4 | NP_056493.3 | ||
| XM_005266346.4 | XP_005266403.1 | |||
| XM_005266347.4 | XP_005266404.2 | |||
| XM_005266348.4 | XP_005266405.2 | |||
| XM_006719803.3 | XP_006719866.1 | |||
| XM_006719805.3 | XP_006719868.1 | |||
| XM_006719806.3 | XP_006719869.1 | |||
| XM_011535046.2 | XP_011533348.1 | |||
| XM_011535047.2 | XP_011533349.1 | |||
| XM_017020544.1 | XP_016876033.1 | |||
| XM_017020545.1 | XP_016876034.1 | |||
| XM_017020546.1 | XP_016876035.1 | |||
| AB046764.1 | BAB13370.1 | |||
| AF467288.1 | AAM53531.1 | |||
| AK091469.1 | ||||
| AK122888.1 | ||||
| AK127791.1 | ||||
| AK297773.1 | ||||
| BX640760.1 | CAE45866.1 |
More Information
Set Membership: |
|

Genomic Map