Assay Details
Target Gene Details
Entrez Gene ID: | 7305 |
Gene Name: | TYRO protein tyrosine kinase binding protein |
Gene Aliases: |
DAP12, KARAP, PLOSL |
Location: |
Chr.19:35904401-35908309 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TYROBP | NM_001173514.1 | NP_001166985.1 | ||
| NM_001173515.1 | NP_001166986.1 | |||
| NM_003332.3 | NP_003323.1 | |||
| NM_198125.2 | NP_937758.1 | |||
| NR_033390.1 | ||||
| AF019562.1 | AAD09436.1 | |||
| AJ010098.1 | CAB52288.1 | |||
| AK290385.1 | ||||
| AY074782.1 | AAL74017.1 | |||
| BC011175.1 | AAH11175.1 | |||
| BI908458.1 | ||||
| BP295666.1 | ||||
| BP302545.1 | ||||
| BP305672.1 | ||||
| BQ576278.1 | ||||
| BT009851.1 | AAP88853.1 | |||
| CR450342.1 | CAG29338.1 | |||
| CR542202.1 | CAG46999.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv51n68 | Chr.19:35841011 - 36016692 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map