Assay Details
Target Gene Details
Entrez Gene ID: | 10257 |
Gene Name: | ATP binding cassette subfamily C member 4 |
Gene Aliases: |
MOAT-B, MOATB, MRP4 |
Location: |
Chr.13:95019829-95301550 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 35 - Exon 35 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCC4 | NM_001301829.1 | NP_001288758.1 | ||
| NM_005845.4 | NP_005836.2 | |||
| XM_005254025.2 | XP_005254082.1 | |||
| XM_017020319.1 | XP_016875808.1 | |||
| XM_017020321.1 | XP_016875810.1 | |||
| AF071202.1 | AAC27076.1 | |||
| AF071203.1 | AAC27077.1 | |||
| AF541977.1 | AAN17334.1 | |||
| AK309460.1 | ||||
| AY081219.1 | AAL88745.1 | |||
| AY207008.1 | AAO37649.1 | |||
| BX647118.1 | ||||
| U83660.1 | AAB71757.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525194 | Chr.13:95020696 - 95084938 on Build GRCh38 | Loss |
|
| nsv832688 | Chr.13:94980939 - 95127944 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map