Assay Details
Target Gene Details
Entrez Gene ID: | 118442 |
Gene Name: | G protein-coupled receptor 62 |
Gene Aliases: |
GPCR8, KPG_005 |
Location: |
Chr.3:51955314-51957504 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR62 | NM_080865.3 | 1 | 1468 | NP_543141.3 |
| AB032600.1 | 1 | 1434 | BAD83591.1 | |
| BC026357.1 | 1 | 1401 | AAH26357.1 | |
| HQ709188.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003453 | Chr.3:51928065 - 52065425 on Build GRCh38 | Gain |
|
| nsv508922 | Chr.3:51940932 - 52009520 on Build GRCh38 | Insertion |
|
| nsv954857 | Chr.3:51932485 - 52056584 on Build GRCh38 | Deletion |
|
| nsv520211 | Chr.3:51738331 - 52132131 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116027996] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map