Assay Details
Target Gene Details
Entrez Gene ID: | 26994 |
Gene Name: | ring finger protein 11 |
Gene Aliases: |
CGI-123, SID1669 |
Location: |
Chr.1:51236273-51273447 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF11 | NM_014372.4 | 3 | 2324 | NP_055187.1 |
| AB024703.1 | 3 | 1967 | BAA84683.1 | |
| AF151881.1 | 3 | 1967 | AAD34118.1 | |
| AL049294.1 | 1 | 1105 | ||
| BC020964.2 | 3 | 1919 | AAH20964.1 | |
| BC047654.1 | 3 | 2292 | AAH47654.1 |
More Information
Set Membership: |
|

Genomic Map