Assay Details
Target Gene Details
Entrez Gene ID: | 55534 |
Gene Name: | mastermind like transcriptional coactivator 3 |
Gene Aliases: |
CAGH3, ERDA3, GDN, MAM-2, MAM2, TNRC3 |
Location: |
Chr.4:139716391-140154079 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAML3 | NM_018717.4 | 5 | 5589 | NP_061187.2 |
| R15711.1 | 1 | 162 |
Target Gene Details
Entrez Gene ID: | 4258 |
Gene Name: | microsomal glutathione S-transferase 2 |
Gene Aliases: |
GST2, MGST-II |
Location: |
Chr.4:139665725-139754546 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGST2 | NM_001204366.1 | NP_001191295.1 | ||
| NM_001204367.1 | NP_001191296.1 | |||
| XM_017008211.1 | XP_016863700.1 | |||
| XM_017008212.1 | XP_016863701.1 | |||
| XM_017008214.1 | XP_016863703.1 | |||
| AK307231.1 | ||||
| DB257575.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3602439 | Chr.4:139692926 - 139951592 on Build GRCh38 | Gain |
|
| esv3602440 | Chr.4:139693390 - 139830109 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115815850] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map