Assay Details
Target Gene Details
Entrez Gene ID: | 5156 |
Gene Name: | platelet derived growth factor receptor alpha |
Gene Aliases: |
CD140A, GAS9, PDGFR-2, PDGFR2, RHEPDGFRA |
Location: |
Chr.4:54229097-54298245 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDGFRA | NM_006206.4 | NP_006197.1 | ||
| XM_005265743.1 | XP_005265800.1 | |||
| XM_006714039.3 | XP_006714102.1 | |||
| XM_006714041.3 | XP_006714104.1 | |||
| XM_011534385.2 | XP_011532687.1 | |||
| XM_017008280.1 | XP_016863769.1 | |||
| XM_017008281.1 | XP_016863770.1 | |||
| XM_017008282.1 | XP_016863771.1 | |||
| AK308353.1 | ||||
| AK311006.1 | ||||
| AK316578.1 | ||||
| BC015186.1 | AAH15186.1 | |||
| BC063414.1 | AAH63414.1 | |||
| DA678599.1 | ||||
| M21574.1 | AAA96715.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1008497 | Chr.4:54249524 - 54261982 on Build GRCh38 | Gain |
|
| dgv5209n100 | Chr.4:54237649 - 54266322 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map