Assay Details
Target Gene Details
Entrez Gene ID: | 84292 |
Gene Name: | WD repeat domain 83 |
Gene Aliases: |
MORG1 |
Location: |
Chr.19:12666804-12675832 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDR83 | NM_001099737.2 | 3 | 362 | NP_001093207.1 |
| NM_032332.3 | 1 | 105 | NP_115708.1 | |
| NR_029375.1 | 3 | 362 | ||
| AK074525.1 | 3 | 352 | ||
| AK223195.1 | 1 | 55 | BAD96915.1 | |
| AK314164.1 | 3 | 339 | ||
| BC005870.2 | AAH05870.1 | |||
| CB143206.1 | 1 | 105 |
Target Gene Details
Entrez Gene ID: | 51398 |
Gene Name: | WD repeat domain 83 opposite strand |
Gene Aliases: |
C19orf56, PTD008 |
Location: |
Chr.19:12668067-12671023 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDR83OS | XM_017026864.1 | 2 | 839 | XP_016882353.1 |
| AK091030.1 | 1 | 1550 | ||
| BG818399.1 | 1 | 129 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953977 | Chr.19:12666887 - 13095486 on Build GRCh38 | Deletion |
|
| nsv1160586 | Chr.19:12597480 - 12823909 on Build GRCh38 | Duplication |
|
| dgv138n111 | Chr.19:12627853 - 12810372 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map