Assay Details
Target Gene Details
Entrez Gene ID: | 10238 |
Gene Name: | DDB1 and CUL4 associated factor 7 |
Gene Aliases: |
AN11, HAN11, SWAN-1, WDR68 |
Location: |
Chr.17:63550435-63594279 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCAF7 | NM_005828.4 | 1 | 42 | NP_005819.3 |
| NR_073585.1 | 1 | 42 | ||
| AK294925.1 | ||||
| AK303212.1 | ||||
| BQ221019.1 | ||||
| DA230140.1 | ||||
| HY023965.1 | 1 | 43 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952355 | Chr.17:63522640 - 63550539 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map