Assay Details
Target Gene Details
Entrez Gene ID: | 3801 |
Gene Name: | kinesin family member C3 |
Gene Aliases: |
- |
Location: |
Chr.16:57758217-57863053 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 30 - Exon 30 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525464 | Chr.16:57714251 - 57832787 on Build GRCh38 | Gain |
|
| nsv471089 | Chr.16:57722731 - 57788355 on Build GRCh38 | Loss |
|
| nsv523237 | Chr.16:57753115 - 57786197 on Build GRCh38 | Loss |
|
| nsv528250 | Chr.16:57715692 - 57786197 on Build GRCh38 | Loss |
|
| nsv509622 | Chr.16:57720104 - 57826484 on Build GRCh38 | Insertion |
|
| nsv952038 | Chr.16:57734189 - 57774488 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map