Assay Details
Target Gene Details
Entrez Gene ID: | 11234 |
Gene Name: | HPS5, biogenesis of lysosomal organelles complex 2 subunit 2 |
Gene Aliases: |
AIBP63, BLOC2S2 |
Location: |
Chr.11:18278670-18322498 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HPS5 | NM_007216.3 | 22 | 3870 | NP_009147.3 |
| NM_181507.1 | 23 | 4027 | NP_852608.1 | |
| NM_181508.1 | 22 | 3683 | NP_852609.1 | |
| XM_011519862.1 | 24 | 4152 | XP_011518164.1 | |
| XM_011519863.1 | 24 | 4060 | XP_011518165.1 | |
| XM_011519864.1 | 24 | 3992 | XP_011518166.1 | |
| XM_011519865.1 | 23 | 4374 | XP_011518167.1 | |
| XM_011519866.1 | 23 | 4025 | XP_011518168.1 | |
| XM_011519867.1 | 23 | 3891 | XP_011518169.1 | |
| XM_011519868.2 | 23 | 4132 | XP_011518170.1 | |
| XM_017017149.1 | 23 | 3926 | XP_016872638.1 | |
| XM_017017150.1 | 23 | 3858 | XP_016872639.1 | |
| XM_017017151.1 | 23 | 4210 | XP_016872640.1 | |
| XM_017017152.1 | 22 | 4240 | XP_016872641.1 | |
| XM_017017153.1 | 22 | 4076 | XP_016872642.1 | |
| XM_017017154.1 | 22 | 3757 | XP_016872643.1 | |
| AB023234.1 | 22 | 3683 | ||
| AF534400.1 | 22 | 3683 | AAO25962.1 | |
| AF534401.1 | 23 | 4027 | AAO25963.1 | |
| AF534402.1 | 22 | 3870 | AAO25964.1 | |
| AJ131721.1 | 11 | 2201 | CAB38232.1 | |
| AK024320.1 | 8 | 1810 | ||
| BC033640.1 | 22 | 3831 | AAH33640.1 |
More Information
Set Membership: |
|

Genomic Map