Assay Details
Target Gene Details
Entrez Gene ID: | 166647 |
Gene Name: | adhesion G protein-coupled receptor A3 |
Gene Aliases: |
GPR125, PGR21, TEM5L |
Location: |
Chr.4:22387374-22516058 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRA3 | NM_145290.3 | 17 | 2804 | NP_660333.2 |
| XM_011513811.2 | 12 | 1988 | XP_011512113.1 | |
| XM_017007834.1 | 17 | 2330 | XP_016863323.1 | |
| AK027494.1 | 6 | 857 | ||
| AK095866.1 | 12 | 1986 | ||
| AK299168.1 | 14 | 2091 | ||
| AL832257.1 | 9 | 1273 | ||
| AY181243.1 | 15 | 2105 | AAO27355.1 | |
| BC035645.1 | 17 | 2815 | AAH35645.1 | |
| BX648287.1 | 1 | 2270 |
More Information
Set Membership: |
|

Genomic Map