Assay Details
Target Gene Details
Entrez Gene ID: | 1643 |
Gene Name: | damage specific DNA binding protein 2 |
Gene Aliases: |
DDBB, UV-DDB2, XPE |
Location: |
Chr.11:47214942-47239218 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDB2 | NM_000107.2 | NP_000098.1 | ||
| NM_001300734.1 | NP_001287663.1 | |||
| AB107037.1 | BAD12557.1 | |||
| AB107038.1 | BAD12558.1 | |||
| AB107039.1 | BAD12559.1 | |||
| AB107040.1 | BAD12560.1 | |||
| AK091640.1 | ||||
| AK313262.1 | ||||
| BC000093.2 | AAH00093.1 | |||
| BT007139.1 | AAP35803.1 | |||
| DB053816.1 | ||||
| U18300.1 | AAB07897.1 |
More Information
Set Membership: |
|

Genomic Map