Assay Details
Target Gene Details
Entrez Gene ID: | 25966 |
Gene Name: | C2 calcium dependent domain containing 2 |
Gene Aliases: |
C21orf25, C21orf258, TMEM24L |
Location: |
Chr.21:41885110-41953957 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C2CD2 | NM_015500.1 | 14 | 6178 | NP_056315.1 |
| NM_199050.2 | 13 | 5677 | NP_950251.1 | |
| XM_011529522.1 | 14 | 6278 | XP_011527824.1 | |
| XM_017028315.1 | 11 | 5414 | XP_016883804.1 | |
| AB047784.1 | 14 | 6178 | BAD74068.1 | |
| AI143200.1 | 1 | 172 | ||
| AI298951.1 | 1 | 172 | ||
| AK125120.1 | 5 | 4747 | ||
| AL050173.1 | 3 | 4392 | CAB43307.1 | |
| CR627455.1 | 6 | 5952 | CAH10536.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv587574 | Chr.21:41877778 - 41893574 on Build GRCh38 | Loss |
|
| dgv7850n54 | Chr.21:41870944 - 41887220 on Build GRCh38 | Loss |
|
| nsv587564 | Chr.21:41833660 - 41887220 on Build GRCh38 | Loss |
|
| dgv7849n54 | Chr.21:41863326 - 41898934 on Build GRCh38 | Loss |
|

Genomic Map