Assay Details
Target Gene Details
Entrez Gene ID: | 64769 |
Gene Name: | MYST/Esa1 associated factor 6 |
Gene Aliases: |
C1orf149, CENP-28, EAF6, NY-SAR-91 |
Location: |
Chr.1:37489960-37514819 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEAF6 | NM_001270875.1 | NP_001257804.1 | ||
| NM_001270876.1 | NP_001257805.1 | |||
| NM_022756.5 | NP_073593.2 | |||
| NR_073090.1 | ||||
| NR_073091.1 | ||||
| NR_073092.1 | ||||
| AK021792.1 | BAB13898.1 | |||
| AK225434.1 | ||||
| AK225598.1 | ||||
| AK308118.1 | ||||
| AK309663.1 | ||||
| AY007166.1 | ||||
| AY211926.1 | AAO65179.1 | |||
| BC016328.1 | AAH16328.1 | |||
| BC056406.1 | AAH56406.1 | |||
| BM840891.1 | ||||
| BX386498.2 | ||||
| BX538212.1 | CAD98071.1 | |||
| BX640719.1 | CAE45838.1 | |||
| DQ099384.1 | AAZ13760.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519551 | Chr.1:37491775 - 37509678 on Build GRCh38 | Gain |
|
| esv2666365 | Chr.1:37507156 - 37524769 on Build GRCh38 | Deletion |
|
| nsv524834 | Chr.1:37496012 - 37585984 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map