Assay Details
Target Gene Details
Entrez Gene ID: | 6301 |
Gene Name: | seryl-tRNA synthetase |
Gene Aliases: |
SERRS, SERS |
Location: |
Chr.1:109213893-109238182 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SARS | NM_006513.3 | 1 | 125 | NP_006504.2 |
| NR_034072.1 | 1 | 125 | ||
| XM_006710813.2 | 1 | 125 | XP_006710876.1 | |
| AK222677.1 | 1 | 86 | BAD96397.1 | |
| AK312771.1 | 1 | 86 | ||
| BC000716.1 | AAH00716.1 | |||
| BC009390.2 | 1 | 72 | AAH09390.1 | |
| BC111398.1 | 1 | 104 | ||
| BP363347.1 | 1 | 125 | ||
| D49914.1 | BAA95602.1 | |||
| DA792457.1 | 1 | 114 | ||
| X91257.1 | 1 | 100 | CAA62635.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1008458 | Chr.1:109097053 - 109231404 on Build GRCh38 | Gain |
|
| nsv1012267 | Chr.1:109168205 - 109279887 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map