Assay Details
Target Gene Details
Entrez Gene ID: | 79447 |
Gene Name: | PAXIP1 associated glutamate rich protein 1 |
Gene Aliases: |
C16orf53, GAS, PA1 |
Location: |
Chr.16:29816207-29822495 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAGR1 | NM_024516.3 | 3 | 957 | NP_078792.1 |
| BC003640.2 | 3 | 653 | AAH03640.1 | |
| EF195235.1 | 3 | 653 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833186 | Chr.16:29719552 - 29863674 on Build GRCh38 | Loss |
|
| nsv827611 | Chr.16:29808186 - 29830422 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map