Assay Details
Target Gene Details
Entrez Gene ID: | 113451 |
Gene Name: | antizyme inhibitor 2 |
Gene Aliases: |
ADC, AZI2, AZIB1, ODC-p, ODC1L, ODCp |
Location: |
Chr.1:33081110-33162288 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AZIN2 | XM_017000167.1 | XP_016855656.1 | ||
| XM_017000174.1 | XP_016855663.1 |
Target Gene Details
Entrez Gene ID: | 55223 |
Gene Name: | tripartite motif containing 62 |
Gene Aliases: |
DEAR1 |
Location: |
Chr.1:33144603-33184774 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIM62 | NM_018207.2 | 5 | 3435 | NP_060677.2 |
| XM_011541705.2 | 5 | 3055 | XP_011540007.1 | |
| XM_017001629.1 | 6 | 3285 | XP_016857118.1 | |
| XM_017001630.1 | 6 | 3369 | XP_016857119.1 | |
| AK001621.1 | 5 | 3031 | BAA91792.1 | |
| AK122896.1 | ||||
| AX775831.1 | 5 | 2839 | CAE11615.1 | |
| BC001222.1 | 5 | 2830 | AAH01222.1 | |
| BC007999.1 | 5 | 2830 | AAH07999.1 | |
| BC011689.2 | 5 | 2830 | AAH11689.1 | |
| BC012152.1 | 5 | 2830 | AAH12152.1 |
More Information
Set Membership: |
|

Genomic Map