Assay Details
Target Gene Details
Entrez Gene ID: | 375061 |
Gene Name: | family with sequence similarity 89 member A |
Gene Aliases: |
C1orf153 |
Location: |
Chr.1:231018958-231040249 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM89A | NM_198552.2 | 2 | 382 | NP_940954.1 |
| BC009447.1 | 2 | 372 | AAH09447.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3589093 | Chr.1:230482037 - 231277190 on Build GRCh38 | Gain |
|
| nsv1000488 | Chr.1:230799658 - 231269735 on Build GRCh38 | Gain |
|
| nsv1011066 | Chr.1:230940432 - 231242850 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map