Assay Details
Target Gene Details
Entrez Gene ID: | 4807 |
Gene Name: | nescient helix-loop-helix 1 |
Gene Aliases: |
HEN1, NSCL, NSCL1, bHLHa35 |
Location: |
Chr.1:160367071-160372848 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NHLH1 | NM_005598.3 | 2 | 1949 | NP_005589.1 |
| BC013789.1 | 2 | 1936 | AAH13789.1 | |
| M96739.1 | 2 | 1951 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548056 | Chr.1:160363952 - 160422364 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map