Assay Details
Target Gene Details
Entrez Gene ID: | 725 |
Gene Name: | complement component 4 binding protein beta |
Gene Aliases: |
C4BP |
Location: |
Chr.1:207088826-207099993 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C4BPB | NM_000716.3 | 3 | 648 | NP_000707.1 |
| NM_001017364.1 | 3 | 645 | NP_001017364.1 | |
| NM_001017365.2 | 4 | 510 | NP_001017365.1 | |
| NM_001017366.2 | 4 | 507 | NP_001017366.1 | |
| NM_001017367.1 | 4 | 484 | NP_001017367.1 | |
| XM_005273254.4 | 5 | 789 | XP_005273311.1 | |
| XM_005273255.2 | 3 | 569 | XP_005273312.1 | |
| AK293379.1 | 3 | 350 | ||
| AK315137.1 | 3 | 569 | ||
| BC005378.1 | 3 | 645 | AAH05378.1 | |
| BG568018.1 | 4 | 510 | ||
| BM987840.1 | ||||
| L11245.1 | 3 | 633 | AAA35616.1 | |
| M29964.1 | 3 | 383 | AAB59520.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832426 | Chr.1:206962184 - 207129471 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map