Assay Details
Target Gene Details
Entrez Gene ID: | 160622 |
Gene Name: | general receptor for phosphoinositides 1 associated scaffold protein |
Gene Aliases: |
TAMALIN |
Location: |
Chr.12:52006945-52015889 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRASP | NM_001271856.1 | NP_001258785.1 | ||
| NM_181711.3 | NP_859062.1 | |||
| XM_005268691.3 | XP_005268748.1 | |||
| XM_011537996.1 | XP_011536298.1 | |||
| BC035500.2 | AAH35500.1 | |||
| BC052967.1 | AAH52967.1 | |||
| BC053666.1 | AAH53666.1 | |||
| HY035281.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2629n54 | Chr.12:52009326 - 52049869 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map