Assay Details
Target Gene Details
Entrez Gene ID: | 196513 |
Gene Name: | decapping mRNA 1B |
Gene Aliases: |
DCP1 |
Location: |
Chr.12:1941591-2004535 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCP1B | NM_152640.4 | 7 | 1337 | NP_689853.3 |
| NR_135060.1 | 8 | 1463 | ||
| XM_011520927.2 | 8 | 1458 | XP_011519229.1 | |
| AK056200.1 | 7 | 1259 | BAB71118.1 | |
| AK299203.1 | 8 | 1398 | ||
| AK310475.1 | 7 | 1128 | ||
| AY146652.1 | 7 | 1313 | AAN62764.1 | |
| BC015368.2 | 3 | 844 | AAH15368.2 | |
| BC043437.2 | 7 | 1275 | AAH43437.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521838 | Chr.12:1951414 - 2190514 on Build GRCh38 | Gain |
|
| nsv1040370 | Chr.12:1912229 - 2561851 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map