Assay Details
Target Gene Details
Entrez Gene ID: | 79812 |
Gene Name: | multimerin 2 |
Gene Aliases: |
EMILIN-3, EMILIN3, ENDOGLYX-1 |
Location: |
Chr.10:86935540-86971311 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MMRN2 | NM_024756.2 | 5 | 690 | NP_079032.2 |
| XM_005270153.1 | 5 | 729 | XP_005270210.1 | |
| XM_006717970.3 | 5 | 2669 | XP_006718033.1 | |
| AK023527.1 | 5 | 637 | BAB14599.1 | |
| AK293822.1 | ||||
| AK304727.1 | 5 | 547 | ||
| AK308401.1 | 4 | 531 | ||
| BC064415.1 | 5 | 645 | AAH64415.1 | |
| BC094744.1 | 5 | 637 | AAH94744.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428236 | Chr.10:86866859 - 87550005 on Build GRCh38 | Loss |
|
| nsv517487 | Chr.10:86899290 - 86973435 on Build GRCh38 | Gain+Loss |
|
| esv2759774 | Chr.10:86755301 - 87550005 on Build GRCh38 | Gain+Loss |
|
| nsv470969 | Chr.10:86936624 - 86972907 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map