Assay Details
Target Gene Details
Entrez Gene ID: | 8835 |
Gene Name: | suppressor of cytokine signaling 2 |
Gene Aliases: |
CIS2, Cish2, SOCS-2, SSI-2, SSI2, STATI2 |
Location: |
Chr.12:93569822-93626236 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOCS2 | NM_003877.4 | 1 | 44 | NP_003868.1 |
| XM_017020147.1 | XP_016875636.1 | |||
| AL522912.3 | 1 | 52 |
Target Gene Details
Entrez Gene ID: | 144481 |
Gene Name: | SOCS2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.12:93565628-93571398 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOCS2-AS1 | NR_038263.1 | |||
| AK054607.1 | ||||
| AK098304.1 |
More Information
Set Membership: |
|

Genomic Map