Assay Details
Target Gene Details
Entrez Gene ID: | 57575 |
Gene Name: | protocadherin 10 |
Gene Aliases: |
OL-PCDH, PCDH19 |
Location: |
Chr.4:133149290-133208606 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDH10 | NM_020815.2 | 1 | 2247 | NP_065866.1 |
| NM_032961.2 | 1 | 2247 | NP_116586.1 | |
| XM_011532150.1 | 1 | 2247 | XP_011530452.1 | |
| AB037821.1 | 1 | 2222 | ||
| AK314377.1 | 1 | 1597 | ||
| AY013874.1 | 1 | 2222 | AAK21987.1 | |
| BC111560.1 | 1 | 1585 | ||
| KJ534913.1 | 1 | 1396 | ||
| KJ534914.1 | 2 | 1228 |
More Information
Set Membership: |
|

Genomic Map