Assay Details
Target Gene Details
Entrez Gene ID: | 11099 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 21 |
Gene Aliases: |
PTPD1, PTPRL10 |
Location: |
Chr.14:88465778-88554779 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPN21 | NM_007039.3 | 19 | 3985 | NP_008970.2 |
| XM_005267287.2 | 19 | 4034 | XP_005267344.1 | |
| XM_006720011.3 | 18 | 3827 | XP_006720074.1 | |
| XM_011536367.2 | 19 | 7153 | XP_011534669.1 | |
| XM_011536368.2 | 17 | 3397 | XP_011534670.1 | |
| XM_011536369.2 | 19 | 3979 | XP_011534671.1 | |
| XM_017020938.1 | 18 | 3936 | XP_016876427.1 | |
| XM_017020939.1 | 18 | 3525 | XP_016876428.1 | |
| AI800682.1 | 1 | 249 | ||
| X79510.1 | 19 | 3986 | CAA56042.1 |
Target Gene Details
Entrez Gene ID: | 55812 |
Gene Name: | spermatogenesis associated 7 |
Gene Aliases: |
HEL-S-296, HSD-3.1, HSD3, LCA3 |
Location: |
Chr.14:88385638-88470350 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPATA7 | XM_006720205.1 | XP_006720268.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv456391 | Chr.14:88427866 - 88573973 on Build GRCh38 | Gain |
|
| esv3635277 | Chr.14:88463565 - 88664404 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map