Assay Details
Target Gene Details
Entrez Gene ID: | 64127 |
Gene Name: | nucleotide binding oligomerization domain containing 2 |
Gene Aliases: |
ACUG, BLAU, CARD15, CD, CLR16.3, IBD1, NLRC2, NOD2B, PSORAS1 |
Location: |
Chr.16:50693581-50733077 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOD2 | NM_022162.2 | NP_071445.1 | ||
| XM_005256084.3 | XP_005256141.1 | |||
| XM_006721242.3 | XP_006721305.1 | |||
| XM_006721243.3 | XP_006721306.1 | |||
| XM_011523260.2 | XP_011521562.1 | |||
| XM_011523261.1 | XP_011521563.1 | |||
| XM_017023536.1 | XP_016879025.1 | |||
| XM_017023537.1 | XP_016879026.1 | |||
| AF178930.1 | AAG33677.1 | |||
| AY187242.1 | ||||
| AY187243.1 | ||||
| AY187244.1 | 1 | 189 | ||
| AY187245.1 | ||||
| AY187246.1 | ||||
| DQ868973.1 | ||||
| HQ204571.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3638608 | Chr.16:50654254 - 50854693 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map