Assay Details
Target Gene Details
Entrez Gene ID: | 144348 |
Gene Name: | zinc finger protein 664 |
Gene Aliases: |
ZFOC1, ZNF176 |
Location: |
Chr.12:123973215-124015439 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF664 | NM_001204298.1 | 2 | 181 | NP_001191227.1 |
| NM_152437.2 | 2 | 230 | NP_689650.1 | |
| AK056034.1 | 2 | 137 | ||
| AL834266.1 | 1 | 855 | CAD38941.1 | |
| BC051696.1 | 2 | 163 | AAH51696.1 | |
| BX647762.1 | 2 | 150 | CAH10567.1 | |
| DA304849.1 | 2 | 230 | ||
| DA328413.1 | 2 | 230 |
Target Gene Details
Entrez Gene ID: | 100533183 |
Gene Name: | filamin-interacting protein FAM101A |
Gene Aliases: |
- |
Location: |
Chr.12:123973215-124316024 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF664-FAM101A | NM_001204299.1 | 2 | 230 | NP_001191228.1 |
| DA169732.1 | 2 | 204 |
More Information
Set Membership: |
|

Genomic Map