Assay Details
Target Gene Details
Entrez Gene ID: | 80021 |
Gene Name: | transmembrane protein 62 |
Gene Aliases: |
- |
Location: |
Chr.15:43133115-43185147 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525461 | Chr.15:42770350 - 43189929 on Build GRCh38 | Loss |
|
| nsv569241 | Chr.15:42908911 - 43230724 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map