Assay Details
Target Gene Details
Entrez Gene ID: | 327 |
Gene Name: | acylaminoacyl-peptide hydrolase |
Gene Aliases: |
AARE, ACPH, APH, D3F15S2, D3S48E, DNF15S2, OPH |
Location: |
Chr.3:49673102-49683508 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APEH | NM_001640.3 | NP_001631.3 | ||
| XM_005265097.2 | XP_005265154.1 | |||
| XM_005265098.4 | XP_005265155.1 | |||
| XM_011533658.2 | XP_011531960.1 | |||
| XM_011533659.1 | XP_011531961.1 | |||
| XM_011533660.2 | XP_011531962.1 | |||
| XM_011533661.2 | XP_011531963.1 | |||
| XM_011533662.2 | XP_011531964.1 | |||
| XM_011533663.2 | XP_011531965.1 | |||
| XM_017006285.1 | XP_016861774.1 | |||
| XM_017006286.1 | XP_016861775.1 | |||
| AK314962.1 | ||||
| BC000362.2 | AAH00362.1 | |||
| BC001499.2 | AAH01499.1 | |||
| BC001826.2 | AAH01826.1 | |||
| D38441.1 | BAA07476.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508217 | Chr.3:49578013 - 49732463 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map