Assay Details
Target Gene Details
Entrez Gene ID: | 28512 |
Gene Name: | NFKB inhibitor interacting Ras like 1 |
Gene Aliases: |
KBRAS1, kappaB-Ras1 |
Location: |
Chr.3:23889536-23917046 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKIRAS1 | XM_005265076.3 | 5 | 2725 | XP_005265133.1 |
| XM_005265077.4 | 4 | 2693 | XP_005265134.1 | |
| XM_005265078.3 | 5 | 2677 | XP_005265135.1 |
Target Gene Details
Entrez Gene ID: | 7324 |
Gene Name: | ubiquitin conjugating enzyme E2 E1 |
Gene Aliases: |
UBCH6 |
Location: |
Chr.3:23805893-23891640 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2E1 | NM_001202476.1 | 5 | 1148 | NP_001189405.1 |
| NM_003341.4 | 6 | 1443 | NP_003332.1 | |
| NM_182666.2 | 5 | 1392 | NP_872607.1 | |
| BC009139.1 | 6 | 1367 | AAH09139.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005186 | Chr.3:21989661 - 24089392 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map