Assay Details
Target Gene Details
Entrez Gene ID: | 10082 |
Gene Name: | glypican 6 |
Gene Aliases: |
OMIMD1 |
Location: |
Chr.13:93226825-94408020 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPC6 | NM_005708.3 | NP_005699.1 | ||
| XM_011521044.2 | XP_011519346.1 | |||
| XM_017020298.1 | XP_016875787.1 | |||
| XM_017020299.1 | XP_016875788.1 | |||
| XM_017020300.1 | XP_016875789.1 | |||
| XM_017020301.1 | XP_016875790.1 | |||
| AF105267.1 | AAD55749.1 | |||
| AF111178.1 | AAD31392.1 | |||
| AK074741.1 | ||||
| AK130013.1 | 3 | 678 | ||
| AK290144.1 | ||||
| AK304338.1 | ||||
| AY358462.1 | AAQ88827.1 | |||
| BC106947.2 | AAI06948.1 |
Target Gene Details
Entrez Gene ID: | 100873973 |
Gene Name: | GPC6 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.13:93818424-93858473 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPC6-AS2 | NR_046536.1 | |||
| BI827445.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892424 | Chr.13:92937463 - 94196357 on Build GRCh38 | Gain |
|
| nsv832684 | Chr.13:93727254 - 93898600 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map