Assay Details
Target Gene Details
Entrez Gene ID: | 80975 |
Gene Name: | transmembrane protease, serine 5 |
Gene Aliases: |
SPINESIN |
Location: |
Chr.11:113687546-113709811 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMPRSS5 | NM_001288749.1 | NP_001275678.1 | ||
| NM_001288750.1 | NP_001275679.1 | |||
| NM_001288751.1 | NP_001275680.1 | |||
| NM_001288752.1 | NP_001275681.1 | |||
| NM_030770.3 | NP_110397.2 | |||
| NR_110046.1 | ||||
| NR_110047.1 | ||||
| XM_017018366.1 | XP_016873855.1 | |||
| XM_017018367.1 | XP_016873856.1 | |||
| AB028140.1 | BAB20375.1 | |||
| AK294313.1 | ||||
| AK295366.1 | ||||
| BC121802.1 | ||||
| BC121803.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3627779 | Chr.11:113688847 - 113692129 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map