Assay Details
Target Gene Details
Entrez Gene ID: | 6752 |
Gene Name: | somatostatin receptor 2 |
Gene Aliases: |
- |
Location: |
Chr.17:73165021-73171955 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SSTR2 | NM_001050.2 | 2 | 1508 | NP_001041.1 |
| AK290745.1 | 2 | 1519 | ||
| AK313866.1 | ||||
| BC000256.1 | 1 | 859 | AAH00256.1 | |
| BC009522.1 | 1 | 1080 | AAH09522.1 | |
| BC019610.1 | 2 | 1508 | AAH19610.1 | |
| BC095495.1 | AAH95495.1 | |||
| BT019926.1 | AAV38729.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833534 | Chr.17:73155642 - 73349355 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map